5-Bromo-2-hydroxybenzophenone, 97%, Thermo Scientific Chemicals
5-Bromo-2-hydroxybenzophenone, 97%, Thermo Scientific Chemicals
5-Bromo-2-hydroxybenzophenone, 97%, Thermo Scientific Chemicals
5-Bromo-2-hydroxybenzophenone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Bromo-2-hydroxybenzophenone, 97%, Thermo Scientific Chemicals
CAS: 55082-33-2 | C13H9BrO2 | 277.117 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ALFB25507.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material5-Bromo-2-hydroxybenzophenone
CAS55082-33-2
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c