2,4-Dichloro-5-methoxyaniline, 98%, Thermo Scientific Chemicals
2,4-Dichloro-5-methoxyaniline, 98%, Thermo Scientific Chemicals
2,4-Dichloro-5-methoxyaniline, 98%, Thermo Scientific Chemicals
2,4-Dichloro-5-methoxyaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,4-Dichloro-5-methoxyaniline, 98%, Thermo Scientific Chemicals
CAS: 98446-49-2 | C7H7Cl2NO | 192.039 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number ALFB25497.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS616-47-7
IUPAC Name1-methyl-1H-imidazole
Molecular FormulaC4H6N2
InChI KeyMCTWTZJPVLRJOU-UHFFFAOYSA-N
SMILESCN1C=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Water=<0.5 % (K.F.)
Appearance (Form)Liquid
AppearanceMay discolor to yellow on storage