2,4-Dichloro-5-methoxyaniline, 98%, Thermo Scientific Chemicals
2,4-Dichloro-5-methoxyaniline, 98%, Thermo Scientific Chemicals
2,4-Dichloro-5-methoxyaniline, 98%, Thermo Scientific Chemicals
2,4-Dichloro-5-methoxyaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,4-Dichloro-5-methoxyaniline, 98%, Thermo Scientific Chemicals
CAS: 98446-49-2 | C7H7Cl2NO | 192.039 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number ALFB25497.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS4105-92-4
IUPAC Name1,3,5-triethyl benzene-1,3,5-tricarboxylate
Molecular FormulaC15H18O6
InChI KeyKXGOWZRHSOJOLF-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CC(=CC(=C1)C(=O)OCC)C(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)129.5-138.5?C
Appearance (Color)White