4-(Methylthio)benzaldehyde, 97%, Thermo Scientific Chemicals
4-(Methylthio)benzaldehyde, 97%, Thermo Scientific Chemicals
4-(Methylthio)benzaldehyde, 97%, Thermo Scientific Chemicals
4-(Methylthio)benzaldehyde, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-(Methylthio)benzaldehyde, 97%, Thermo Scientific Chemicals
CAS: 3446-89-7 | C8H8OS | 152.211 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFB25427.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material4-(Methylthio)benzaldehyde
CAS3446-89-7
Health Hazard 1H302
Health Hazard 3P264b-P270-P301+P312-P330-P501c
Melting Point6°C
View more
4-(Methylthio)benzaldehyde is a building block for the synthesis of various pharmaceutical and biologically active compounds. It can be used for the synthesis of sulfur-containing terpyridine ligands. It is the intermediate for the synthesis of a class of pyrrole derivatives showing analgesic/anti-inflammatory activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-(Methylthio)benzaldehyde is a building block for the synthesis of various pharmaceutical and biologically active compounds. It can be used for the synthesis of sulfur-containing terpyridine ligands. It is the intermediate for the synthesis of a class of pyrrole derivatives showing analgesic/anti-inflammatory activity.
Solubility
Not miscible in water.
Notes
Air sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Ara Nunez-Montenegroa; Rosa Carballoa; Jose M.Hermida-Ramonb; Ezequiel M.Vazquez-Lopez. Synthesis, characterization, reactivity and computational studies of new rhenium(I) complexes with thiosemicarbazone ligands derived from 4-(methylthio)benzaldehyde. Polyhedron. 2011, 30,(12), 2146-2156.
  2. Piotr Filipiak; Gordon L.Hug; Krzysztof Bobrowski; Bronislaw Marciniak. Photochemistry of 4-(methylthio)phenylacetic acid: Steady-state and laser flash photolysis studies. Journal of Photochemistry and Photobiology A: Chemistry. 2005, 172,(3), 322-330.