3-Heptene, cis + trans, 94%, Thermo Scientific™
3-Heptene, cis + trans, 94%, Thermo Scientific™
3-Heptene, cis + trans, 94%, Thermo Scientific™
3-Heptene, cis + trans, 94%, Thermo Scientific™
3-Heptene, cis + trans, 94%, Thermo Scientific™
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFB25297.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material3-Heptene
Name Notecis + trans
CAS592-78-9
Health Hazard 1H225
Health Hazard 2GHS H Statement
H225
Highly flammable liquid and vapour.
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.