Chlorpheniramine maleate, 99%, Thermo Scientific Chemicals
Chlorpheniramine maleate, 99%, Thermo Scientific Chemicals
Chlorpheniramine maleate, 99%, Thermo Scientific Chemicals
Chlorpheniramine maleate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chlorpheniramine maleate, 99%, Thermo Scientific Chemicals
CAS: 113-92-8 | C20H23ClN2O4 | 390.86 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFB25238.14
Price (MYR)
663.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
663.00
Add to cart
Specifications
Chemical Name or MaterialChlorpheniramine maleate
CAS113-92-8
Health Hazard 1H302
Health Hazard 2GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3P264b-P270-P301+P312-P330-P501c
View more
An antihistaminic agent