4-(4-Hydroxy-3-methoxyphenyl)-2-butanone, 97%, Thermo Scientific Chemicals
4-(4-Hydroxy-3-methoxyphenyl)-2-butanone, 97%, Thermo Scientific Chemicals
4-(4-Hydroxy-3-methoxyphenyl)-2-butanone, 97%, Thermo Scientific Chemicals
4-(4-Hydroxy-3-methoxyphenyl)-2-butanone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-(4-Hydroxy-3-methoxyphenyl)-2-butanone, 97%, Thermo Scientific Chemicals
CAS: 122-48-5 | C11H14O3 | 194.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
Catalog number ALFB25150.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS88574-06-5
IUPAC Name6-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
Molecular FormulaC21H23NO4
InChI KeyFPCPONSZWYDXRD-UHFFFAOYSA-N
SMILESOC(=O)CCCCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%