4-(4-Hydroxy-3-methoxyphenyl)-2-butanone, 97%, Thermo Scientific Chemicals
4-(4-Hydroxy-3-methoxyphenyl)-2-butanone, 97%, Thermo Scientific Chemicals
4-(4-Hydroxy-3-methoxyphenyl)-2-butanone, 97%, Thermo Scientific Chemicals
4-(4-Hydroxy-3-methoxyphenyl)-2-butanone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-(4-Hydroxy-3-methoxyphenyl)-2-butanone, 97%, Thermo Scientific Chemicals
CAS: 122-48-5 | C11H14O3 | 194.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
25 g
Catalog number ALFB25150.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material4-(4-Hydroxy-3-methoxyphenyl)-2-butanone
CAS122-48-5
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.