DL-Panthenol, 99%, Thermo Scientific Chemicals
DL-Panthenol, 99%, Thermo Scientific Chemicals
DL-Panthenol, 99%, Thermo Scientific Chemicals
DL-Panthenol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
DL-Panthenol, 99%, Thermo Scientific Chemicals
CAS: 16485-10-2 | C9H19NO4 | 205.25 g/mol
Have Questions?
Catalog NumberQuantity
ALFB25114.1850 g
Catalog number ALFB25114.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS2110-18-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
FormLiquid
Assay (GC)≥98.5%
Refractive Index1.5585-1.5615 @ 20?C
DL-Panthenol is used in enantiospecific HPLC separation of pantothenic acid, panthenol and pantolactone enantiomers. It promotes stimulation of epithelization, granulation, mitigation of itching and has an anti-inflammatory effect. It is effective for moisturization, improvement of hair structure and adding luster and sheen to the hair.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
DL-Panthenol is used in enantiospecific HPLC separation of pantothenic acid, panthenol and pantolactone enantiomers. It promotes stimulation of epithelization, granulation, mitigation of itching and has an anti-inflammatory effect. It is effective for moisturization, improvement of hair structure and adding luster and sheen to the hair.
Solubility
Soluble in water, ethanol, ether, chloroform, and propylene glycol.
Notes
Hygroscopic. Store away from moisture and oxidizing agents under dry inert gas. Protect from humidity and water. Incompatible with oxidizing agents and moisture.
RUO – Research Use Only
General References:
  1. Takashi Arai; Hiroyasu Matsuda; Hiroshi Ōizumi. Determination of optical purity by high-performance liquid chromatography on chiral stationary phases: pantothenic acid and related compounds. Journal of Chromatography A.1989, 474, (5), 405-410.
  2. Morton. Schmall; E. G. Wollish. Determination of Panthenol and Pantothenates in Multivitamin Preparations. Anal. Chem.1957, 29, (10), 1509-1513.