3-Fluorophenyl isothiocyanate, 97+%, Thermo Scientific Chemicals
3-Fluorophenyl isothiocyanate, 97+%, Thermo Scientific Chemicals
3-Fluorophenyl isothiocyanate, 97+%, Thermo Scientific Chemicals
3-Fluorophenyl isothiocyanate, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Fluorophenyl isothiocyanate, 97+%, Thermo Scientific Chemicals
CAS: 404-72-8 | C7H4FNS | 153.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ALFB25001.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS2305-25-1
IUPAC Nameethyl 3-hydroxyhexanoate
Molecular FormulaC8H16O3
InChI KeyLYRIITRHDCNUHV-UHFFFAOYNA-N
SMILESCCCC(O)CC(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive Index1.4265-1.4315 @ 20?C
FormLiquid
Assay (GC)≥96.0%