Iodocyclohexane, 98%, stab. with copper, Thermo Scientific Chemicals
Iodocyclohexane, 98%, stab. with copper, Thermo Scientific Chemicals
Iodocyclohexane, 98%, stab. with copper, Thermo Scientific Chemicals
Iodocyclohexane, 98%, stab. with copper, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Iodocyclohexane, 98%, stab. with copper, Thermo Scientific Chemicals
CAS: 626-62-0 | C6H11I | 210.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number ALFB24840.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS14064-21-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to dark red-brown
Assay (GC)≥97.5%
FormLiquid
Refractive Index1.5765-1.5805 @ 20°C
Iodocyclohexane has been employed as reagent in demethylation of aryl methyl ethers in DMF under reflux condition.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Iodocyclohexane has been employed as reagent in demethylation of aryl methyl ethers in DMF under reflux condition.
Solubility
Soluble in ethanol, ether, acetone.
Notes
Light sensitive. Store away from oxidizing agent, light.
RUO – Research Use Only
General References:
  1. D K Zaouris; A M Wenge; D Murdock; T A A Oliver; G Richmond; G A D Ritchie; R N Dixon; M N R Ashfold. Conformer specific dissociation dynamics of iodocyclohexane studied by velocity map imaging. Journal of Chemical Physics. 2011, 135 (9), 094312.
  2. Jun Yi; Xi Lu; Yan-Yan Sun; Bin Xiao; Lei Liu. Nickel-catalyzed sonogashira reactions of non-activated secondary alkyl bromides and iodides. Angewandte Chemie. International edition in English. 2013, 52 (47), 12409-12413.