Aminoguanidine hydrogen carbonate, 98+%, Thermo Scientific Chemicals
Aminoguanidine hydrogen carbonate, 98+%, Thermo Scientific Chemicals
Aminoguanidine hydrogen carbonate, 98+%, Thermo Scientific Chemicals
Aminoguanidine hydrogen carbonate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Aminoguanidine hydrogen carbonate, 98+%, Thermo Scientific Chemicals
CAS: 2582-30-1 | C2H8N4O3 | 136.111 g/mol
Have Questions?
Catalog NumberQuantity
ALFB24696.22100 g
Catalog number ALFB24696.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAminoguanidine hydrogen carbonate
CAS2582-30-1
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H317
May cause an allergic skin reaction.
Health Hazard 3GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
View more
Aminoguanidine bicarbonate is an inhibitor of NOS (nitric oxide synthase). Aminoguanidine bicarbonate is used to study the effect of addition of polyamines to rat embryo cell cultures infected with adenovirus type 5.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Aminoguanidine bicarbonate is an inhibitor of NOS (nitric oxide synthase). Aminoguanidine bicarbonate is used to study the effect of addition of polyamines to rat embryo cell cultures infected with adenovirus type 5.
Solubility
Soluble in water (3.3 mg/ml at 30°C), and water (2.7 mg/ml at 20°C).
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
General References:
  1. T. George.; C. L. Kaul.; R. S. Grewal.; R. Tahilramani. Antihypertensive and monoamine oxidize inhibitory activity of some derivatives of 3-formyl-4-oxo-4H-pyrido[1,2-a]pyrimidine. J. Med. Chem. 1971, 14, (10), 913-915.
  2. Jaroslav Stanek.; Giorgio Caravatti.; Jorg Frei.; Pascal Furet.; Helmut Mett.; Peter Schneider.; Urs Regenass. 4-Amidinoindan-1-one 2'-amidinohydrazone: a new potent and selective inhibitor of S-adenosylmethionine decarboxylase. J. Med. Chem. 1993, 36, (15), 2168-2171.