1H,1H,2H,2H-Perfluorooctyltriethoxysilane, 97%, Thermo Scientific Chemicals
1H,1H,2H,2H-Perfluorooctyltriethoxysilane, 97%, Thermo Scientific Chemicals
1H,1H,2H,2H-Perfluorooctyltriethoxysilane, 97%, Thermo Scientific Chemicals
1H,1H,2H,2H-Perfluorooctyltriethoxysilane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1H,1H,2H,2H-Perfluorooctyltriethoxysilane, 97%, Thermo Scientific Chemicals
CAS: 51851-37-7 | C14H19F13O3Si | 510.367 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFB24620.14
Price (MYR)
2,779.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
2,779.00
Add to cart
Specifications
Boiling Point220°C
Chemical Name or Material1H,1H,2H,2H-Perfluorooctyltriethoxysilane
Melting Point<-38°C
CAS51851-37-7
Health Hazard 1Warning
View more
1H,1H,2H,2H-Perfluorooctyltriethoxysilane is used as an active component of fluorine modified ORMOCER (organically modified ceramics) coating materials. It is a fluorine based polymer having low surface free energy, enhances its anti-adhesive behavior of polar and non-polar substances. Its wetting property acts in repelling aqueous electrolyte solution away from metallic substrates, thereby giving protection to metal substrate from corrosion. It is utilized in the surface modification of glass nanoparticles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1H,1H,2H,2H-Perfluorooctyltriethoxysilane is used as an active component of fluorine modified ORMOCER (organically modified ceramics) coating materials. It is a fluorine based polymer having low surface free energy, enhances its anti-adhesive behavior of polar and non-polar substances. Its wetting property acts in repelling aqueous electrolyte solution away from metallic substrates, thereby giving protection to metal substrate from corrosion. It is utilized in the surface modification of glass nanoparticles.
Solubility
Miscible with alcohol, aromatic and aliphatic hydrocarbons.
Notes
Moisture sensitive. Incompatible with moisture and oxidizing agents.
RUO – Research Use Only
General References:
  1. Kujawa, J.; Cerneaux, S.; Kujawski, W. Highly hydrophobic ceramic membranes applied to the removal of volatile organic compounds in pervaporation. Chem. Eng. J. 2015, 260, 43-54.
  2. Bahng, J. H.; Yeom, B.; Wang, Y.; Tung, S. O.; Hoff, J. D.; Kotov, N. Anomalous dispersions of/hedgehog/'particles. Nature 2015, 517 (7536), 596-599.