1-(4-Nitrophenylazo)-2-naphthol, Thermo Scientific Chemicals
1-(4-Nitrophenylazo)-2-naphthol, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-(4-Nitrophenylazo)-2-naphthol, Thermo Scientific Chemicals
CAS: 6410-10-2 | C16H11N3O3 | 293.28 g/mol
Have Questions?
Catalog NumberQuantity
ALFB24600.1425 g
Catalog number ALFB24600.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS372-39-4
IUPAC Name3,5-difluoroaniline
Molecular FormulaC6H5F2N
InChI KeyKQOIBXZRCYFZSO-UHFFFAOYSA-N
SMILESNC1=CC(F)=CC(F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
AppearanceMay darken on storage
Appearance (Color)White to yellow
Infrared spectrumConforms
GC>=97.5 %
Appearance (Form)Low melting solid