2-Aminopyrimidine, 98%, Thermo Scientific Chemicals
2-Aminopyrimidine, 98%, Thermo Scientific Chemicals
2-Aminopyrimidine, 98%, Thermo Scientific Chemicals
2-Aminopyrimidine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Aminopyrimidine, 98%, Thermo Scientific Chemicals
CAS: 109-12-6 | C4H5N3 | 95.11 g/mol
Have Questions?
Catalog NumberQuantity
ALFB24594.22100 g
Catalog number ALFB24594.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS34590-94-8
IUPAC Name2-(2-methoxypropoxy)propan-1-ol
Molecular FormulaC7H16O3
InChI KeyCUDYYMUUJHLCGZ-UHFFFAOYNA-N
SMILESCOC(C)COC(C)CO
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 % sum of isomers
Infrared spectrumConforms
Appearance (Color)Clear colorless
Appearance (Form)Liquid