1-Ethyl-2-pyrrolidinone, 98%, Thermo Scientific Chemicals
1-Ethyl-2-pyrrolidinone, 98%, Thermo Scientific Chemicals
1-Ethyl-2-pyrrolidinone, 98%, Thermo Scientific Chemicals
1-Ethyl-2-pyrrolidinone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Ethyl-2-pyrrolidinone, 98%, Thermo Scientific Chemicals
CAS: 2687-91-4 | C6H11NO | 113.16 g/mol
Have Questions?
Catalog NumberQuantity
ALFB24548.36500 g
Catalog number ALFB24548.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Ethyl-2-pyrrolidinone
CAS2687-91-4
Health Hazard 1H227-H318-H360Df
Health Hazard 2GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3P201-P202-P210-P281-P305+P351+P338-P308+P313-P310-P370+P378q-P501c