2-Aminoterephthalic acid, 99%, Thermo Scientific Chemicals
2-Aminoterephthalic acid, 99%, Thermo Scientific Chemicals
2-Aminoterephthalic acid, 99%, Thermo Scientific Chemicals
2-Aminoterephthalic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Aminoterephthalic acid, 99%, Thermo Scientific Chemicals
CAS: 10312-55-7 | C8H5NO4 | 179.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ALFB24441.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS458-45-7
IUPAC Name3-(3-fluorophenyl)propanoic acid
Molecular FormulaC9H9FO2
InChI KeyUBLMRADOKLXLCD-UHFFFAOYSA-N
SMILESOC(=O)CCC1=CC(F)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Appearance (Color)White to cream or pale yellow
Melting Point (clear melt)39.0-48.0°C
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥96.0%
2-Aminoterephthalic acid is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Aminoterephthalic acid is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Polya M Miladinova and Temenushka N Konstantinova. Synthesis and properties of some new blue-emitting triazine derivatives of 2-aminoterephthalic acid, containing a polymerisable group and stabiliser fragment. Coloration Technology. 2009, 125 (4), 242-247.
  2. J. Arias-Pardilla; H.J. Salavagione; C. Barbero; E. Morallón; J.L. Vázquez. Study of the chemical copolymerization of 2-aminoterephthalic acid and aniline.: Synthesis and copolymer properties. European Polymer Journal. 2006, 42 (7), 1521-1532.