beta-Naphthol Violet, Thermo Scientific™
beta-Naphthol Violet, Thermo Scientific™
beta-Naphthol Violet, Thermo Scientific™
beta-Naphthol Violet, Thermo Scientific™
beta-Naphthol Violet, Thermo Scientific™
Have Questions?
Catalog NumberQuantity
ALFB24279.0910 g
Catalog number ALFB24279.09
Price (MYR)
68.96
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (MYR)
68.96
Add to cart
Specifications
Chemical Name or Materialbeta-Naphthol Violet
CAS7143-21-7
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Beta-Naphthol violet exhibits antiseptic properties and is used in the synthesis of certain azo dyes and antioxidants for rubber. It can also be used in the detection of the ferric ion which turns the Beta-Naphthol violet solution green.
Solubility
Soluble in water.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
General References:
  1. Thomas E. Archer; James D. Stokes. Residue analysis of .beta.-naphthoxyacetic acid and .beta.-naphthol by high-pressure liquid chromatography. J. Agric. Food Chem. 1978, 26 (2), 452-455.
  2. William J. Husa and Lydia M. Husa. The effect of various compounds on the rate of development of rancidity in fats and oils. Journal of the American Pharmaceutical Association. 1928, 17 (3), 243-247.