1-Octylamine, 99%, Thermo Scientific Chemicals
1-Octylamine, 99%, Thermo Scientific Chemicals
1-Octylamine, 99%, Thermo Scientific Chemicals
1-Octylamine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Octylamine, 99%, Thermo Scientific Chemicals
CAS: 111-86-4 | C8H19N | 129.247 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFB24193.36
Price (MYR)
621.00
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
621.00
Add to cart
Specifications
Chemical Name or Material1-Octylamine
CAS111-86-4
Health Hazard 1H226-H301+H311-H314-H332-H335
Health Hazard 2GHS H Statement
H314-H318-H302-H312-H332-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Combustible liquid.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
View more
1-Octylamine is used as a precursor to prepare chemicals. It finds application in functional fluids and as a laboratory reagent. Further, it is used as a reactant in the preparation of 2H-indazoles and 1H-indazolones, which are used as myeloperoxidase(MPO) inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Octylamine is used as a precursor to prepare chemicals. It finds application in functional fluids and as a laboratory reagent. Further, it is used as a reactant in the preparation of 2H-indazoles and 1H-indazolones, which are used as myeloperoxidase(MPO) inhibitors.
Solubility
Miscible with ethanol, benzene and ethyl acetate. Slightly miscible with water.
Notes
Incompatible with acids, strong oxidizing agents, acid anhydrides and acid chlorides.
RUO – Research Use Only
General References:
  1. Firmansyah, D.; Banasiewicz, M.; Gryko, D. T. Vertically-expanded imidazo[1,2-a]pyridines and imidazo[1,5-a]pyridinevia dehydrogenative coupling. Org. Biomol. Chem. 2015, 13 (5), 1367-1374.
  2. Liu, Z.; Chiang, C. Y.; Li, W.; Zhou, W. The role of surface hydrolysis of ferricyanide anions in crystal growth of snowflake-shaped alpha-Fe2O3. Chem. Commun. 2015, 51 (45), 9350-9353.