Maltitol, 97%, Thermo Scientific Chemicals
Maltitol, 97%, Thermo Scientific Chemicals
Maltitol, 97%, Thermo Scientific Chemicals
Maltitol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Maltitol, 97%, Thermo Scientific Chemicals
CAS: 585-88-6 | C12H24O11 | 344.313 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number ALFB24158.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS15283-45-1
IUPAC Namegold(1+) trisodium disulfanidesulfonate
Molecular FormulaAuNa3O6S4
InChI KeyFJCMCHNDCIYVST-UHFFFAOYSA-J
SMILES[Na+].[Na+].[Na+].[Au+].[O-]S([S-])(=O)=O.[O-]S([S-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (special)Au: ≥34.8%
FormPowder
Total Metal Impurities≤0.1%
Maltitol is used in the manufacturing of sweets, including sugarless hard candies, chewing gum, chocolates, baked goods and ice cream. In the pharmaceutical industry, it is used as an excipient. Further, it is used as a plasticizer in gelatin capsules, humectant, emollient and a low-calorie sweetening agent. It is involved in the study to investigate the comparative effects of the maltitol chewing gums on reducing plaque.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Maltitol is used in the manufacturing of sweets, including sugarless hard candies, chewing gum, chocolates, baked goods and ice cream. In the pharmaceutical industry, it is used as an excipient. Further, it is used as a plasticizer in gelatin capsules, humectant, emollient and a low-calorie sweetening agent. It is involved in the study to investigate the comparative effects of the maltitol chewing gums on reducing plaque.
Solubility
Soluble in water. Slightly soluble in ethanol.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Konar, N.; Poyrazoglu, E. S.; Artik, N. Influence of calcium fortification on physical and rheological properties of sucrose-free prebiotic milk chocolates containing inulin and maltitol. J. Food Sci. Technol. 2015, 52 (4), 2033-2042.
  2. Chen, M.; Wu, S.; Tang, W.; Gong, J. Caking and adhesion free energy of maltitol: Studying of mechanism in adhesion process. Powder Technol. 2015, 272, 235-240.