3,4-Dihydroxybenzoic acid, 97%, Thermo Scientific Chemicals
3,4-Dihydroxybenzoic acid, 97%, Thermo Scientific Chemicals
3,4-Dihydroxybenzoic acid, 97%, Thermo Scientific Chemicals
3,4-Dihydroxybenzoic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3,4-Dihydroxybenzoic acid, 97%, Thermo Scientific Chemicals
CAS: 99-50-3 | C7H6O4 | 154.121 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFB24016.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS7558-79-4
IUPAC Namedisodium hydrogen phosphate
Molecular FormulaHNa2O4P
InChI KeyBNIILDVGGAEEIG-UHFFFAOYSA-L
SMILES[Na+].[Na+].OP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
pH6.98 to 7.02 (at 20°C) composition: 3.5 g
pHpotassium dihydrogen phosphate and 12.5 g
pHdi-sodium hydrogen phosphate per liter
Appearance (Form)Solution
3,4-Dihydroxybenzoic acid is a metabolite of polyphenols such as phloretin and quercetin. It is also a phenolic acid antioxidant present in fruits, vegetables and nuts. Further, it is found to be an efficacious chemopreventive agent in several carcinogenesis models. In addition to this, it acts as an anti-inflammatory agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,4-Dihydroxybenzoic acid is a metabolite of polyphenols such as phloretin and quercetin. It is also a phenolic acid antioxidant present in fruits, vegetables and nuts. Further, it is found to be an efficacious chemopreventive agent in several carcinogenesis models. In addition to this, it acts as an anti-inflammatory agent.
Solubility
Soluble in hot alcohol and ether. Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only
General References:
  1. Chandra, I. K.; Angkawijaya, A. E.; Santoso, S. P.; Ismadji, S.; Soetaredjo, F. E.; Ju, Y. H. Solution equilibria studies of complexes of divalent metal ions with 2-aminophenol and 3,4-dihydroxybenzoic acid. Polyhedron 2015, 88, 29-39.
  2. Tang, Y.; Li, X.; Zhang, B.; Chen, P. X.; Liu, R.; Tsao, R. Characterisation of phenolics, betanins and antioxidant activities in seeds of three Chenopodium quinoa Willd. genotypes. Food Chem. 2015, 166, 308-388.