4-(Diethylamino)azobenzene, 98%, Thermo Scientific Chemicals
4-(Diethylamino)azobenzene, 98%, Thermo Scientific Chemicals
4-(Diethylamino)azobenzene, 98%, Thermo Scientific Chemicals
4-(Diethylamino)azobenzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-(Diethylamino)azobenzene, 98%, Thermo Scientific Chemicals
CAS: 2481-94-9 | C16H19N3 | 253.349 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFB23915.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS161321-36-4
IUPAC Name2-cyclohexyl-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)acetic acid
Molecular FormulaC23H25NO4
InChI KeyBWQQGHPODCJZDB-UHFFFAOYNA-N
SMILESOC(=O)C(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%