4-(Diethylamino)azobenzene, 98%, Thermo Scientific Chemicals
4-(Diethylamino)azobenzene, 98%, Thermo Scientific Chemicals
4-(Diethylamino)azobenzene, 98%, Thermo Scientific Chemicals
4-(Diethylamino)azobenzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-(Diethylamino)azobenzene, 98%, Thermo Scientific Chemicals
CAS: 2481-94-9 | C16H19N3 | 253.349 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number ALFB23915.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material4-(Diethylamino)azobenzene
CAS2481-94-9
Health Hazard 1H302-H315-H317-H319
Health Hazard 2GHS H Statement
H341
Suspected of causing genetic defects.
Health Hazard 3P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P330-P333+P313-P362-P501c