1-Methylpyrrolidine, 98%, Thermo Scientific Chemicals
1-Methylpyrrolidine, 98%, Thermo Scientific Chemicals
1-Methylpyrrolidine, 98%, Thermo Scientific Chemicals
1-Methylpyrrolidine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Methylpyrrolidine, 98%, Thermo Scientific Chemicals
CAS: 120-94-5 | C5H11N | 85.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ALFB23799.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Methylpyrrolidine
CAS120-94-5
Health Hazard 1H225-H301-H314-H332-H335
Health Hazard 2GHS H Statement
H225-H314-H318-H302-H332
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
View more
1-Methylpyrrolidine is a methylated pyrollidine and is involved as an essential part of the structure of cefipime. It is also an active component of cigarette smoke.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Methylpyrrolidine is a methylated pyrollidine and is involved as an essential part of the structure of cefipime. It is also an active component of cigarette smoke.
Solubility
Miscible with water.
Notes
Air sensitive. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only
General References:
  1. Pohlmann, S.; Olyschlager, T.; Goodrich, P.; Vicente, J. A.; Jacquemin, J.; Balducci, A. Azepanium-based ionic liquids as green electrolytes for high voltage supercapacitors. J. Power Sources 2015, 273, 931-936.
  2. Dabrowska, M.; Starek, M.; Krzek, J.; Papp, E.; Krol, P. A degradation study of cefepime hydrochloride in solutions under various stress conditions by TLC-densitometry. Biomed. Chromatogr. 2015, 29 (3), 388-395.