2-(Dimethylamino)ethanol, 99+%, Thermo Scientific Chemicals
2-(Dimethylamino)ethanol, 99+%, Thermo Scientific Chemicals
2-(Dimethylamino)ethanol, 99+%, Thermo Scientific Chemicals
2-(Dimethylamino)ethanol, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-(Dimethylamino)ethanol, 99+%, Thermo Scientific Chemicals
CAS: 108-01-0 | C4H11NO | 89.138 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 mL
250 mL
Catalog number ALFB23616.AU
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1000 mL
Request bulk or custom format
Chemical Identifiers
CAS7681-65-4
IUPAC Nameλ¹-copper(1+) iodide
Molecular FormulaCuI
InChI KeyLSXDOTMGLUJQCM-UHFFFAOYSA-M
SMILES[Cu+].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofA≥32.5% Cu
Appearance (Color)White to pale brown or grey
FormPowder
Assay from Suppliers CofA≥97.5% (Complexometric titration)
2-(Dimethylamino)ethanol is used as corrosion inhibitor, anti-scaling agent, paint additive, coating additive and solids separation agent. It is also used as an intermediate for active pharmaceutical ingredients and dyes. It serves as a curing agent for polyurethanes and epoxy resins. Further, it is used as an additive to boiler water. In addition to this, it is used therapeutically as a CNS stimulant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Miscible with water, alcohol, ether, acetone and benzene.
Notes
Incompatible with strong oxidizers and strong acids.
RUO – Research Use Only
General References:
  1. In the presence of 2 mol of n-BuLi, forms a unimetal superbase complex of the Li alkoxide with an additional molecule of n-BuLi: J. Chem. Soc., Perkin 1, 3071 (1997). For examples of its use, see 2-Methoxypyridine, A13675, 2-Chloropyridine, A14866.
  2. Hadri, N. E. L.; Quang, D. V.; Abu-Zahra, M. R. M. Study of Novel Solvent for CO2 Post-combustion Capture. Energy Procedia 2015, 75, 2268-2286.
  3. Bringas, G.; Navarro-Santos, P.; López-Rendón, R.; López-Lemus, J.; Bresme, F. Molecular Dynamics Simulations of 2-(Dimethylamino)ethanol. J. Phys. Chem. B 2015, 119 (15), 5035-5046.