2-(Methylamino)pyridine, 98%, Thermo Scientific Chemicals
2-(Methylamino)pyridine, 98%, Thermo Scientific Chemicals
2-(Methylamino)pyridine, 98%, Thermo Scientific Chemicals
2-(Methylamino)pyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-(Methylamino)pyridine, 98%, Thermo Scientific Chemicals
CAS: 4597-87-9 | C6H8N2 | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFB23589.14
Price (MYR)
1,419.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
1,419.00
Add to cart
Specifications
Chemical Name or Material2-(Methylamino)pyridine
CAS4597-87-9
Health Hazard 1H227-H302+H312+H332-H315-H319-H335
Health Hazard 2GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c