(+)-Griseofulvin, 97%, Thermo Scientific Chemicals
(+)-Griseofulvin, 97%, Thermo Scientific Chemicals
(+)-Griseofulvin, 97%, Thermo Scientific Chemicals
(+)-Griseofulvin, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
(+)-Griseofulvin, 97%, Thermo Scientific Chemicals
CAS: 126-07-8 | C17H17ClO6 | 352.767 g/mol
Have Questions?
Catalog NumberQuantity
ALFB23521.1425 g
Catalog number ALFB23521.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS25704-18-1
Molecular Formula(C8H7NaO3S)n
SMILES[Na]OS(=O)(=O)C1=CC=C(C=C1)C(-*)C-*
MDL NumberMFCD00084449
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)White to tan
Infrared spectrumConforms
Griseofulvin is an antifungal drug. It is used both in animal and in humans, to treat ringworm infections of the skin and nails.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Griseofulvin is an antifungal drug. It is used both in animal and in humans, to treat ringworm infections of the skin and nails.
Solubility
Soluble in dimethyl formamide, ethanol, methanol, chloroform, DMSO, tetrachloroethane. Insoluble in water.
Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
RUO – Research Use Only
General References:
  1. Klaus Weber; Jürgen Wehland; Wilhelm Herzog. Griseofulvin interacts with microtubules both in vivo and in vitro. Journal of Molecular Biology. 1976, 102, (4), 817-829.
  2. Win Loung Chiou and Sidney Riegelman. Preparation and dissolution characteristics of several fast-release solid dispersions of griseofulvin. Journal of Pharmaceutical Sciences. 1969, 58 (12), 1505-1510.