4-(Aminomethyl)benzoic acid, 97%, Thermo Scientific Chemicals
4-(Aminomethyl)benzoic acid, 97%, Thermo Scientific Chemicals
4-(Aminomethyl)benzoic acid, 97%, Thermo Scientific Chemicals
4-(Aminomethyl)benzoic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-(Aminomethyl)benzoic acid, 97%, Thermo Scientific Chemicals
CAS: 56-91-7 | C8H9NO2 | 151.165 g/mol
Have Questions?
Catalog NumberQuantity
ALFB23519.1425 g
Catalog number ALFB23519.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material4-(Aminomethyl)benzoic acid
CAS56-91-7
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
4-(Aminomethyl)benzoic acid acts as an unnatural amino acid derivative. It is also used as a type 2 antifibrinolytic agent. Further, it reacts with 2-methyl-isothiourea sulfate to prepare 4-guanidinomethylbenzoic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-(Aminomethyl)benzoic acid acts as an unnatural amino acid derivative. It is also used as a type 2 antifibrinolytic agent. Further, it reacts with 2-methyl-isothiourea sulfate to prepare 4-guanidinomethylbenzoic acid.
Solubility
Slightly soluble in water. Insoluble in ethanol, benzene and chloroform.
Notes
Incompatible with strong oxidizing agents, strong reducing agents and strong bases.
RUO – Research Use Only
General References:
  1. Li, Z.; Fang, L.; Wang, J.; Dong, L.; Guo, Y.; Xie, Y. An Improved and Practical Synthesis of Tranexamic Acid. Org. Process Res. Dev. 2015, 19 (3), 444-448.
  2. Li, K. S.; Xiao, P.; Zhang, D. I.; Hou, X. B.; Ge, L.; Yang, D. X.; Liu, H. D.; He, D. F.; Chen, X.; Han, K. R.; Song, X. Y.; Yu, X.; Fang, H.; Sun, J. P. Identification of para-Substituted Benzoic Acid Derivatives as Potent Inhibitors of the Protein Phosphatase Slingshot. ChemMedChem 2015, 10 (12), 1980-1987.