Dichloroisocyanuric acid sodium salt, 97% (dry wt.), Thermo Scientific Chemicals
Dichloroisocyanuric acid sodium salt, 97% (dry wt.), Thermo Scientific Chemicals
Dichloroisocyanuric acid sodium salt, 97% (dry wt.), Thermo Scientific Chemicals
Dichloroisocyanuric acid sodium salt, 97% (dry wt.), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dichloroisocyanuric acid sodium salt, 97% (dry wt.), Thermo Scientific Chemicals
CAS: 2893-78-9 | C3HCl2N3NaO3 | 220.949 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFB23504.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS92-86-4
IUPAC Name4,4'-dibromo-1,1'-biphenyl
Molecular FormulaC12H8Br2
InChI KeyHQJQYILBCQPYBI-UHFFFAOYSA-N
SMILESBrC1=CC=C(C=C1)C1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to light beige
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Crystalline powder or crystals
Melting point165°C to 169°C
Sodium dichloroisocyanurate can be used as a reagent for N-monochlorination and dehydrochlorination of amino esters. It is also a reagent for chlorination to detect ammonium via formation of colored zebra-bands in a detecting tube.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium dichloroisocyanurate can be used as a reagent for N-monochlorination and dehydrochlorination of amino esters, synthesis of antimicrobial polyurethane additives to control biofilm. It is used as a emergency disinfectant in drinking water, widely used cleansing agent. It is also a reagent for chlorination to detect ammonium via formation of colored zebra-bands in a detecting tube.
Solubility
Soluble in water.
Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
RUO – Research Use Only
General References:
  1. Tatsuaki Hori; Keizou Niki; Yoshiaki Kiso; Tatsuo Oguchi; Yuki Kamimoto; Toshiro Yamada; Masahiro Nagai; Tatsuaki Hori; Keizou Niki; Yoshiaki Kiso; Tatsuo Oguchi; Yuki Kamimoto; Toshiro Yamada; Masahiro Nagai. Ammonium detection by formation of colored zebra-bands in a detecting tube. Talanta. 2010, 81, (4-5), 1467-1471.
  2. S Devin McLennan; Lauren A Peterson; Joan B Rose. Comparison of point-of-use technologies for emergency disinfection of sewage-contaminated drinking water. Applied and Environmental Microbiology. 2009, 75 (22), 7283-7286.
  3. Reagent for N-chlorination of primary and secondary amines, particularly useful for chlorination of hindered amines: Synth. Commun., 18, 2135 (1988).