Hexabromobenzene, 97%, Thermo Scientific Chemicals
Hexabromobenzene, 97%, Thermo Scientific Chemicals
Hexabromobenzene, 97%, Thermo Scientific Chemicals
Hexabromobenzene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hexabromobenzene, 97%, Thermo Scientific Chemicals
CAS: 87-82-1 | C6Br6 | 551.49 g/mol
Have Questions?
Catalog NumberQuantity
ALFB23295.1425 g
Catalog number ALFB23295.14
Price (MYR)
304.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
304.00
Add to cart
Specifications
Chemical Name or MaterialHexabromobenzene
CAS87-82-1
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c