Hexanoyl chloride, 97%, Thermo Scientific Chemicals
Hexanoyl chloride, 97%, Thermo Scientific Chemicals
Hexanoyl chloride, 97%, Thermo Scientific Chemicals
Hexanoyl chloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Hexanoyl chloride, 97%, Thermo Scientific Chemicals
CAS: 142-61-0 | C6H11ClO | 134.603 g/mol
Have Questions?
Catalog NumberQuantity
ALFB23027.22100 g
Catalog number ALFB23027.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS3483-12-3
IUPAC Name(2R,3R)-1,4-disulfanylbutane-2,3-diol
Molecular FormulaC4H10O2S2
InChI KeyVHJLVAABSRFDPM-IMJSIDKUSA-N
SMILESO[C@@H](CS)[C@@H](O)CS
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Loss on Drying≤0.5% (US sourced material)
Melting Point (clear melt)37.0-45.0?C
View more
Hexanoyl chloride has been used in the synthesis of (±)-7-butyl-6,8-dihydroxy-3-pentyl-3,4-dihydroisochromen-1-one, 14-methyl-1-octadecene. It is used as crystal liquid, pharmaceutical and organic synthesis material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Hexanoyl chloride has been used in the synthesis of (±)-7-butyl-6,8-dihydroxy-3-pentyl-3,4-dihydroisochromen-1-one, 14-methyl-1-octadecene. It is used as crystal liquid, pharmaceutical and organic synthesis material.
Solubility
Soluble in ether, chloroform.
Notes
Moisture sensitive. Store under inert gas. Incompatible with oxidizing agents, water/ moisture.
RUO – Research Use Only
General References:
  1. Aamer Saeed; Hummera Rafique; Muhammad Arshad. Total synthesis and antibacterial screening of ( ± )-7-butyl-6,8-dihydroxy-3-pentyl-3,4-dihydroisochromen-1-one. Journal of Asian Natural Products Research. 2013, 15 (10), 1112-1122.
  2. Quanjing Zhu; Xiaofeng Chu; Zhaoyan Zhang; Wei-Lin Dai; Kangnian Fan. Effect of the tungsten precursor on the high activity of the WO3/ZrO2 catalyst in the oxidative lactonization of 1,2-benzenedimethanol. Molecules. 2013, 18, (5), 5201-5208.