Phenyltrimethylammonium chloride, 98+%, Thermo Scientific Chemicals
Phenyltrimethylammonium chloride, 98+%, Thermo Scientific Chemicals
Phenyltrimethylammonium chloride, 98+%, Thermo Scientific Chemicals
Phenyltrimethylammonium chloride, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenyltrimethylammonium chloride, 98+%, Thermo Scientific Chemicals
CAS: 138-24-9 | C9H14ClN | 171.67 g/mol
Have Questions?
Catalog NumberQuantity
ALFB22851.22100 g
Catalog number ALFB22851.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS2595-97-3
IUPAC Name(3R,4R,5S,6R)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol
Molecular FormulaC6H12O6
InChI KeyWQZGKKKJIJFFOK-IVMDWMLBSA-N
SMILESOC[C@H]1OC(O)[C@H](O)[C@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥97.0%
Water Content (Karl Fischer Titration)≤1.0%
Optical Rotation+15.0? ? 1.0? (c=5 in Distilled water)
Phenyltrimethylammonium chloride is an effective phase transfer catalyst, methylating agent, also useful for alkaline depolymerizations, corrosion inhibitor for low carbon steel.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phenyltrimethylammonium chloride is an effective phase transfer catalyst, methylating agent, also useful for alkaline depolymerizations, corrosion inhibitor for low carbon steel.
Solubility
Soluble in water 333 g/L.
Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agent, water/ moisture.
RUO – Research Use Only
General References:
  1. Paul G. Sears; Eugene D. Wilhoit and Lyle R. Dawson. Conductances of Trimethylphenylammonium Chloride and Iodide in Water and in Dimethylformamide at 25°C. J. Chem. Phys. 1955, 23 (7), 1274.
  2. T. Jiang; M.J. Chollier Brym; G. Dubéb, A. Lasia; G.M. Brisard. Electrodeposition of Aluminum from ionic liquids: Part II - studies on the electrodeposition of aluminum from aluminum chloride (AICl3) - trimethylphenylammonium chloride (TMPAC) ionic liquids. Surface and Coatings Technology. 2006, 201, (1-2), 10-18.