2-Mercapto-5-nitrobenzimidazole, 97%, Thermo Scientific Chemicals
2-Mercapto-5-nitrobenzimidazole, 97%, Thermo Scientific Chemicals
2-Mercapto-5-nitrobenzimidazole, 97%, Thermo Scientific Chemicals
2-Mercapto-5-nitrobenzimidazole, 97%, Thermo Scientific Chemicals
2-Mercapto-5-nitrobenzimidazole, 97%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFB22358.065 g
Catalog number ALFB22358.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS18039-90-2
IUPAC Nametris(oxolane); trichlorotitanium
Molecular FormulaC12H24Cl3O3Ti
InChI KeyMIDYWIPTCYRMQF-UHFFFAOYSA-K
SMILESCl[Ti](Cl)Cl.C1CCOC1.C1CCOC1.C1CCOC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Blue-green to blue
Titration Argentometric>=96.0 %
Appearance (Form)Powder or crystals