Iodonitrotetrazolium violet, 95%
Iodonitrotetrazolium violet, 95%
Iodonitrotetrazolium violet, 95%
Iodonitrotetrazolium violet, 95%
Thermo Scientific Chemicals
Iodonitrotetrazolium violet, 95%
CAS: 146-68-9 | C19H13ClIN5O2 | 505.70 g/mol
Have Questions?
Catalog NumberdimQuantity
ALFB22301.03
Catalog number ALFB22301.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Specifications
Formula Weight505.70 (Anhydrous)
Unit Size
Used for dehydrogenase determination