Filter aid, Celite Standard Super-cel, Thermo Scientific Chemicals
Filter aid, Celite Standard Super-cel, Thermo Scientific Chemicals
Filter aid, Celite Standard Super-cel, Thermo Scientific Chemicals
Filter aid, Celite Standard Super-cel, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Filter aid, Celite Standard Super-cel, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFB21983.36500 g
Catalog number ALFB21983.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS81-11-8
IUPAC Name5-amino-2-[(1E)-2-(4-amino-2-sulfophenyl)ethenyl]benzene-1-sulfonic acid
Molecular FormulaC14H14N2O6S2
InChI KeyREJHVSOVQBJEBF-OWOJBTEDSA-N
SMILESNC1=CC=C(\C=C\C2=CC=C(N)C=C2S(O)(=O)=O)C(=C1)S(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to yellow-brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with NaOH>=94 %
Filter aid, Celite Standard Super-cel is used to maximize biodiesel yields as a result of improved filtration, dewatering, dewaxing and winterization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Filter aid, Celite Standard Super-cel is used to maximize biodiesel yields as a result of improved filtration, dewatering, dewaxing and winterization.
Solubility
Insoluble in water.
Notes
Incompatible with acids.
RUO – Research Use Only
General References:
  1. Farmani, J. Modeling of solid fat content of chemically interesterified fully hydrogenated soybean oil and canola oil blends as a function of temperature and saturated fatty acids. Journal of Food Measurement and Characterization 2015, 9 (3), 281-289.
  2. Garderen, N. V.; Otal, E. H.; Aneziris, C. G.; Graule, T.; Clemens, F. J. Influence of porous substrate on copper based oxygen carrier efficiency for chemical-looping combustion. Microporous Mesoporous Mater. 2014, 190, 362-370.