4'-Hydroxybiphenyl-4-carboxylic acid, 99%, Thermo Scientific Chemicals
4'-Hydroxybiphenyl-4-carboxylic acid, 99%, Thermo Scientific Chemicals
4'-Hydroxybiphenyl-4-carboxylic acid, 99%, Thermo Scientific Chemicals
4'-Hydroxybiphenyl-4-carboxylic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4'-Hydroxybiphenyl-4-carboxylic acid, 99%, Thermo Scientific Chemicals
CAS: 58574-03-1 | C13H10O3 | 214.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2.5 g
Catalog number ALFB21946.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS145-50-6
IUPAC Name(4E)-4-[(4-hydroxynaphthalen-1-yl)(phenyl)methylidene]-1,4-dihydronaphthalen-1-one
Molecular FormulaC27H18O2
InChI KeyVDDWRTZCUJCDJM-SOYKGTTHSA-N
SMILESOC1=C2C=CC=CC2=C(C=C1)C(\C1=CC=CC=C1)=C1/C=CC(=O)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark red to red-brown to brown-black
Appearance (Color)to orange-brown
Appearance (Form)Powder
Infrared spectrumConforms
Molar extinction coefficient>=50000 (0.01 g/l methanol) (207-213 nm)