4-Iodoaniline, 98%, Thermo Scientific Chemicals
4-Iodoaniline, 98%, Thermo Scientific Chemicals
4-Iodoaniline, 98%, Thermo Scientific Chemicals
4-Iodoaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Iodoaniline, 98%, Thermo Scientific Chemicals
CAS: 540-37-4 | C6H6IN | 219.025 g/mol
Have Questions?
Catalog NumberQuantity
ALFB21936.1850 g
Catalog number ALFB21936.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Iodoaniline
CAS540-37-4
Health Hazard 1H302+H312+H332-H315-H319-H335
Health Hazard 2GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
4-dehydroanilinium ion, a stable distonic isomer of ionized aniline, by collision-activated dissociation of protonated 4-iodoaniline. Palladium-catalyzed carbonylation of 4-iodoaniline gave the corresponding four- armed star polybenzamides. 4-Iodoaniline is a iodine substituted aniline that is widely used as chemical intermediates in the manufacturing of dyes and drugs. 4-Iodoaniline was used to prepare phenyl functionalized graphene oxide (I-Ph-GO).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-dehydroanilinium ion, a stable distonic isomer of ionized aniline, by collision-activated dissociation of protonated 4-iodoaniline. Palladium-catalyzed carbonylation of 4-iodoaniline gave the corresponding four- armed star polybenzamides. 4-Iodoaniline is a iodine substituted aniline that is widely used as chemical intermediates in the manufacturing of dyes and drugs. 4-Iodoaniline was used to prepare phenyl functionalized graphene oxide (I-Ph-GO).
Solubility
Slightly soluble in water.Soluble in chloroform, methanol.
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
RUO – Research Use Only
General References:
  1. Veronica R. Reichert,; Lon J. Mathias.Tetrahedrally-Oriented Four-Armed Star and Branched Aramids. Macromolecules,. 1994, 27 (24), 7024-7029.
  2. Mihály Kádár,; Zoltán Nagy,; Tamás Karancsi,; György Farsang.The electrochemical oxidation of 4-bromoaniline, 2,4-dibromoaniline, 2,4,6-tribromoaniline and 4-iodoaniline in acetonitrile solution. Electrochimica Acta. 2001, 46 22, 3405-3414.