Crystal Violet
Crystal Violet
Crystal Violet
Thermo Scientific Chemicals
Crystal Violet
CAS: 548-62-9 | C25H30ClN3 | 407.986 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number ALFB21932.36
Price (MYR)
1,732.00
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
1,732.00
Add to cart
Specifications
Chemical Name or MaterialCrystal Violet
Melting Point∼215°C (decomposition)
CAS548-62-9
Health Hazard 1H302-H318-H351
Health Hazard 2GHS H Statement
H318-H351-H302
Causes serious eye damage.
Suspected of causing cancer.
Harmful if swallowed.
View more
Crystal Violet is used as an acid-base indicator, alcohol denaturant, biological stain, textile dye, and an indicator for copper salts. It is a component of gram staining in microorganisms and cell lines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Crystal Violet is used as an acid-base indicator, alcohol denaturant, biological stain, textile dye, and an indicator for copper salts. It is a component of gram staining in microorganisms and cell lines.
Solubility
Miscible with water and chloroform. Partially miscible with alcohol and glycerol.
Notes
Incompatible with acids, oxidizing agents, alkali metals, strong oxidizing agents, ammonia, acid chlorides, acid anhydrides, reducing agents and peroxides.
RUO – Research Use Only
General References:
  1. Tsai, H.; Lai, C.; Hsiao, C.; Medina, H.; Su, T.; Ouyang, H.; Chen, T.; Liang, H.; Chueh, Y. Plasma-Assisted Synthesis of High-Mobility Atomically Layered Violet Phosphorus. ACS Appl. Mater. Interfaces 2015, 7 (25), 13723-13727.
  2. Petelenz, P.; Snamina, M.; Mazur, G. Charge-Transfer States in Pentacene: Dimer versusCrystal. J. Phys. Chem. C 2015, 119 (25), 14338-14342.