Digoxin, 96%
Digoxin, 96%
Digoxin, 96%
Thermo Scientific Chemicals
Digoxin, 96%
CAS: 20830-75-5 | C41H64O14 | 780.95 g/mol
Have Questions?
Catalog NumberQuantity
ALFB21902.1425 g
Catalog number ALFB21902.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS6334-96-9
IUPAC Name1-chloro-4-(4-chlorobutoxy)butane
Molecular FormulaC8H16Cl2O
InChI KeyPVBMXMKIKMJQRK-UHFFFAOYSA-N
SMILESClCCCCOCCCCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥98.5%
Refractive Index1.4570-1.4600 @ 20?C
Digoxin is used in several cardiac pathologies such as atrial fibrillation and heart failure.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Digoxin is used in several cardiac pathologies such as atrial fibrillation and heart failure.
Solubility
Soluble in dimethyl sulfoxide and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only