Digoxin, 96%, Thermo Scientific Chemicals
Digoxin, 96%, Thermo Scientific Chemicals
Digoxin, 96%, Thermo Scientific Chemicals
Digoxin, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Digoxin, 96%, Thermo Scientific Chemicals
CAS: 20830-75-5 | C41H64O14 | 780.949 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number ALFB21902.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS7631-86-9
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder
Total Metal Impurities≤0.005% (50 ppm)
Appearance (Color)White
Assay (special)?2.5 % of Theoretical value (Si)
Digoxin is used in several cardiac pathologies such as atrial fibrillation and heart failure.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Digoxin is used in several cardiac pathologies such as atrial fibrillation and heart failure.
Solubility
Soluble in dimethyl sulfoxide and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only
General References:
  1. Barry F. Uretsky et.al. Randomized study assessing the effect of digoxin withdrawal in patients with mild to moderate chronic congestive heart failure: Results of the Proved trial. Journal of the American College of Cardiology., 1993, 22 (4), 955-962.
  2. Rodney H. Falk, M.D.; Anne A. Knowlton, M.D.; Sheilah A. Bernard, M.D.; Norman E. Gotlieb, M.D.; And Nancy J. Battinelli, R.N. Digoxin for Converting Recent-Onset Atrial Fibrillation to Sinus Rhythm: A Randomized, Double-Blinded Trial. Ann Intern Med, 1987, 106 (4), 503-506.