11-Bromo-1-undecene, 90+%, Thermo Scientific Chemicals
11-Bromo-1-undecene, 90+%, Thermo Scientific Chemicals
11-Bromo-1-undecene, 90+%, Thermo Scientific Chemicals
11-Bromo-1-undecene, 90+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
11-Bromo-1-undecene, 90+%, Thermo Scientific Chemicals
CAS: 7766-50-9 | C11H21Br | 233.193 g/mol
Have Questions?
Catalog NumberQuantity
ALFB21849.065 g
Catalog number ALFB21849.06
Price (MYR)
481.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
481.00
Add to cart
Specifications
Chemical Name or Material11-Bromo-1-undecene
CAS7766-50-9
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
11-Bromo-1-undecene is used in advanced polymer dielectrics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
11-Bromo-1-undecene is used in advanced polymer dielectrics.
Solubility
Soluble in water, 0.2422 mg/L @ 25°C (est).
Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Oĝuz Türünç; Lucas Montero de Espinosa1 and Michael A. R. Meier. TRenewable Polyethylene Mimics Derived from Castor Oil. Macromolecular Rapid Communications. 2011, 32 (17), 1357-1361.
  2. Laura J. Yeager; Dafni G. Amirsakis; Elan Newman; Robin L. Garrell. Synthesis of epoxy-terminated dialkyl disulfides for polymerizable self-assembled monolayers. Tetrahedron Letters. 1998, 39 (46), 8409-8412.