3,4-(Methylenedioxy)benzylamine, 97%, Thermo Scientific™
3,4-(Methylenedioxy)benzylamine, 97%, Thermo Scientific™
3,4-(Methylenedioxy)benzylamine, 97%, Thermo Scientific™
Have Questions?
Catalog NumberQuantity
ALFB21837.1425 g
Catalog number ALFB21837.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material3,4-(Methylenedioxy)benzylamine
CAS2620-50-0
Health Hazard 1H314-H335
Health Hazard 2GHS H Statement
H314
Causes severe skin burns and eye damage.
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Piperonylamine is a metabolite of dopamine analogs. It is a pharmaceutical intermediate.
Solubility
Sparingly soluble in water. (0.9 g/100 mL).
Notes
Air sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store at 2-8°C.
General References:
  1. David E. Nichols.; William K. Brewster.; Michael P. Johnson.; Robert Oberlender.; Robert M. Riggs. Nonneurotoxic tetralin and indan analogs of 3,4-(methylenedioxy)amphetamine (MDA). J. Med. Chem. 1990, 33 (2), 703-710.
  2. Nobuhisa Watanabe.; Yasuhiro Kabasawa.; Yasutaka Takase.; Masayuki Matsukura.; Kazuki Miyazaki.; Hiroki Ishihara.; Kohtarou Kodama.; Hideyuki Adachi. 4-Benzylamino-1-chloro-6-substituted Phthalazines: Synthesis and Inhibitory Activity toward Phosphodiesterase 5.J. Med. Chem. 1998, 41 (18),3367-3372.