Tartronic acid, 97%, Thermo Scientific Chemicals
Tartronic acid, 97%, Thermo Scientific Chemicals
Tartronic acid, 97%, Thermo Scientific Chemicals
Tartronic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tartronic acid, 97%, Thermo Scientific Chemicals
CAS: 80-69-3 | C3H4O5 | 120.06 g/mol
Have Questions?
Catalog NumberQuantity
ALFB21640.020.5 g
Catalog number ALFB21640.02
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
0.5 g
Request bulk or custom format
Chemical Identifiers
CAS26311-45-5
IUPAC Name4-pentylbenzoic acid
Molecular FormulaC12H16O2
InChI KeyCWYNKKGQJYAHQG-UHFFFAOYSA-N
SMILESCCCCCC1=CC=C(C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Melting Point (clear melt)84.0-91.0?C
FormPowder
Assay (Silylated GC)≥98.5%
Tartronic acid is used as a reactant in the catalytic oxidation with air to form mesoxalic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tartronic acid is used as a reactant in the catalytic oxidation with air to form mesoxalic acid.
Solubility
Soluble in water and alcohol.
Notes
Store in a cool place. Incompatible with bases, oxidizing agents and reducing agents.
RUO – Research Use Only
General References:
  1. Gomes, J. F.; Garcia, A. C.; Ferreira, E. B.; Pires, C.; Oliveira, V. L.; Tremiliosi-Filho, G.; Gasparotto, L. H. New insights into the formation mechanism of Ag, Au and AgAu nanoparticles in aqueous alkaline media: alkoxides from alcohols, aldehydes and ketones as universal reducing agents. Phys. Chem. Chem. Phys. 2015, 17 (33), 21683-21693.
  2. Wang, H.; Thia, L.; Li, N.; Ge, X.; Liu, Z.; Wang, X. Selective electro-oxidation of glycerol over Au supported on extended poly(4-vinylpyridine) functionalized graphene. Appl. Catal., B 2015, 166, 25-31.