1-Bromo-2,3-difluorobenzene, 98%, Thermo Scientific Chemicals
1-Bromo-2,3-difluorobenzene, 98%, Thermo Scientific Chemicals
1-Bromo-2,3-difluorobenzene, 98%, Thermo Scientific Chemicals
1-Bromo-2,3-difluorobenzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Bromo-2,3-difluorobenzene, 98%, Thermo Scientific Chemicals
CAS: 38573-88-5 | C6H3BrF2 | 192.991 g/mol
Have Questions?
Catalog NumberQuantity
ALFB21585.065 g
Catalog number ALFB21585.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS3326-32-7
IUPAC Name3',6'-dihydroxy-5-isothiocyanato-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
Molecular FormulaC21H11NO5S
InChI KeyMHMNJMPURVTYEJ-UHFFFAOYSA-N
SMILESOC1=CC=C2C(OC3=CC(O)=CC=C3C22OC(=O)C3=CC(=CC=C23)N=C=S)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=88 %
Appearance (Form)Crystalline powder
Appearance (Color)Orange