Diethyl terephthalate, 95%, Thermo Scientific Chemicals
Diethyl terephthalate, 95%, Thermo Scientific Chemicals
Diethyl terephthalate, 95%, Thermo Scientific Chemicals
Diethyl terephthalate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diethyl terephthalate, 95%, Thermo Scientific Chemicals
CAS: 636-09-9 | C12H14O4 | 222.24 g/mol
Have Questions?
Catalog NumberQuantity
ALFB21401.1850 g
Catalog number ALFB21401.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS114615-82-6
IUPAC Nametetrapropylazanium tetrahydrate ruthenium
Molecular FormulaC12H36NO4Ru
InChI KeyHUCLFLGLPCVDMZ-UHFFFAOYSA-N
SMILESO.O.O.O.[Ru].CCC[N+](CCC)(CCC)CCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Black
Ruthenium (Ru)>=27.7 %
Appearance (Form)Powder