2-Acetamidophenol, 97%, Thermo Scientific Chemicals
2-Acetamidophenol, 97%, Thermo Scientific Chemicals
2-Acetamidophenol, 97%, Thermo Scientific Chemicals
2-Acetamidophenol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Acetamidophenol, 97%, Thermo Scientific Chemicals
CAS: 614-80-2 | C8H9NO2 | 151.165 g/mol
Have Questions?
Catalog NumberQuantity
ALFB21264.22100 g
Catalog number ALFB21264.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS142-31-4
IUPAC Namesodium octyl sulfate
Molecular FormulaC8H17NaO4S
InChI KeyWFRKJMRGXGWHBM-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCCOS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Chloride content≤0.1%
Assay from Supplier's CofA≥98.5 to ≤101.5%
Identification (FTIR)Conforms
Appearance (Color)White to pale cream