1,12-Dibromododecane, 98%, Thermo Scientific Chemicals
1,12-Dibromododecane, 98%, Thermo Scientific Chemicals
1,12-Dibromododecane, 98%, Thermo Scientific Chemicals
1,12-Dibromododecane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,12-Dibromododecane, 98%, Thermo Scientific Chemicals
CAS: 3344-70-5 | C12H24Br2 | 328.132 g/mol
Have Questions?
Catalog NumberQuantity
ALFB21254.1425 g
Catalog number ALFB21254.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS13116-27-3
IUPAC Name(4-iodophenyl)hydrazine
Molecular FormulaC6H7IN2
InChI KeyIQMLCMKMSBMMGR-UHFFFAOYSA-N
SMILESNNC1=CC=C(I)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to orange to brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point99°C to 106°C
NMR>=94 %
1,12-Dibromododecane, is used as an organic chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,12-Dibromododecane, is used as an organic chemical intermediate.
Solubility
Soluble in hot methanol- almost transparency.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions.Stable. Combustible. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. R Zana.; M Benrraou.; R Rueff. Alkanediyl-.alpha.,.μ.-bis(dimethylalkylammonium bromide) surfactants. 1. Effect of the spacer chain length on the critical micelle concentration and micelle ionization degree. Langmuir. 19917 (6), 1072-1075.