Guanidine thiocyanate, 99%
Guanidine thiocyanate, 99%
Guanidine thiocyanate, 99%
Guanidine thiocyanate, 99%
Thermo Scientific Chemicals
Guanidine thiocyanate, 99%
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2500 g
Catalog number ALFB21250.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS13472-35-0
IUPAC Namesodium dihydrate dihydrogen phosphate
Molecular FormulaH6NaO6P
InChI KeyVBJGJHBYWREJQD-UHFFFAOYSA-M
SMILESO.O.[Na+].OP(O)([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Adhering crystals
Appearance (Color)White
Titration with NaOH>=99.0 %
Loss on drying=<24 %
Heavy metals (as Pb)=<10 ppm
View more
Guanidine thiocyanate is a useful tool for RNA isolation and protein solubilization. It is involved in the quantification of mRNA from hepatocytes. It acts as a medium for blood smaple lystae in order to maintain the integrity of nucleic acid. It is a chaotropic agent, mainly involved in the extraction of DNA and RNA. It is utilized to deactivate the influenza virus.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Guanidine thiocyanate is a useful tool for RNA isolation and protein solubilization. It is involved in the quantification of mRNA from hepatocytes. It acts as a medium for blood smaple lystae in order to maintain the integrity of nucleic acid. It is a chaotropic agent, mainly involved in the extraction of DNA and RNA. It is utilized to deactivate the influenza virus.
Solubility
Soluble in water and ethanol
Notes
Light sensitive. Incompatible with acids and strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Chomczynski, P.; Sacchi, N. The single-step method of RNA isolation by acid guanidinium thiocyanate-phenol-chloroform extraction: twenty-something years on. Nat. Protoc. 2006, 1 (2), 581-585.
  2. van Doorn, H. R.; Bruijnesteijn van Coppenraet, E. S.; Duim, B.; Vandenbroucke-Grauls, C. M.; Weel, J. F.; Dankert, J.; Kuijper, E. J.; de Jong, M. D. Silica-guanidinium thiocyanate-based nucleic acid isolation protocol does not improve sensitivity of two commercial tests for detection of Mycobacterium tuberculosis in respiratory samples. Eur. J. Clin. Microbiol. Infect. Dis. 2006, 25 (10), 673-675.
Contents & Storage
Ambient temperatures