2-Acetylbenzo[b]thiophene, 98%, Thermo Scientific Chemicals
2-Acetylbenzo[b]thiophene, 98%, Thermo Scientific Chemicals
2-Acetylbenzo[b]thiophene, 98%, Thermo Scientific Chemicals
2-Acetylbenzo[b]thiophene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Acetylbenzo[b]thiophene, 98%, Thermo Scientific Chemicals
CAS: 22720-75-8 | C10H8OS | 176.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number ALFB21173.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS9037-24-5
Molecular FormulaMFCD00145841
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Beads
Total weight capacity>=4.70 eq/kg (H)
Water=<1.5 %
Particle size=<1.0 % (< 0.300 mm)