Giemsa Stain
Giemsa Stain
Giemsa Stain
Thermo Scientific Chemicals
Giemsa Stain
CAS: 51811-82-6 | C14H14ClN3S | 291.80 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number ALFB21172.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS89-93-0
IUPAC Name1-(2-methylphenyl)methanamine
Molecular FormulaC8H11N
InChI KeyCJAAPVQEZPAQNI-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1CN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥95.0%
Refractive Index1.5420-1.5460 @ 20°C
Giemsa Stain is used as a laboratory reagent and stain for microscopy. It is also used for the histopathologic detection of malaria and other parasites such as spirochete and protozoan. The stain is also been incorporated in differential staining between bacterial and human cells. The compound has been used in combination with May-Grunwald or Wright Stain (sc-203763) as a histological hematology stain.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Giemsa Stain is used as a laboratory reagent and stain for microscopy. It is also used for the histopathologic detection of malaria and other parasites such as spirochete and protozoan. The stain is also been incorporated in differential staining between bacterial and human cells. The compound has been used in combination with May-Grunwald or Wright Stain (sc-203763) as a histological hematology stain.
Solubility
Insoluble in water (<0.1%), methanol (5.8 mg/L), and ethanol.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. K. K. Friend.; S. Chen.; F. H. Ruddle. Differential staining of interspecific chromosomes in somatic cell hybrids by alkaline Giemsa stain. Somatic Cell Genetics. 1976, 2 (2), 183-188.
  2. Julie R. Korenberg.; Elizabeth F. Freedlender. Giemsa technique for the detection of sister chromatid exchanges. Ind. Eng. Chem. Res. 1974, 48 (4), 355-360.